Learn More
Hecht Karl™ Glass Balls
Product Code. 10073681
Change view
Click to view available options
Diameter (Metric):
1 mm
2 mm
4 mm
Unit Size:
1kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10073681 4 mm 1kg
10439411 1 mm 1kg
10359111 2 mm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10073681 Supplier Hecht Karl™ Supplier No. 41401004
Estimated Shipment: 07-12-2026
Made of clear, polished glass. Karl Hecht™ Glass Balls can be used as boiling stones, mixing beads or for distillation columns.
 Spherical, solid, polished glass beads for general laboratory use.
Specifications
Type Ball
Material Glass
Diameter (Metric) 4 mm
Quantity 1kg Pack