Learn More
Ansell™ Gladiator™ 16-650 Series Heavy Weight Gloves
Description
- Crinkle finish for grip on slipper surfaces and protection from sharp edges
- Natural rubber coating
- Cut-and-sewn construction
- Interlock cotton liner
- Fully-coated finish
- Gauntlet cuff
Specifications
Specifications
| Coating Material | Natural Rubber Latex |
| Color | Green |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Latex Free | No |
| Material | Rubber Latex |
| Product Type | Heavyweight Gloves |
| Size | 10 |
| Finger Style | Crinkled finish |
| For Use With (Application) | Glass repair and replacement Construction Utilities All food processing applications that have food contact Seafood processing |
| Show More |