missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Givinostat hydrochloride monohydrate, MedChemExpress
Description
Givinostat hydrochloride monohydrate (ITF-2357 hydrochloride monohydrate) is a HDAC inhibitor with an IC50 of 198 and 157 nM for HDAC1 and HDAC3, respectively.
Specifications
Specifications
| Chemical Name or Material | Givinostat hydrochloride monohydrate |
| Physical Form | Solid |
| CAS | 732302-99-7 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30ClN3O5 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | ITF-2357 hydrochloride monohydrate |
| Solubility Information | DMSO : ≥ 100 mg/mL (210.10 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?