Learn More
Gilson™ PIPETMANTM Universal Reload Box
Product Code. 10122862 Shop All Gilson Products
Change view
Click to view available options
Color:
Red, Green, Yellow, Blue
Holds:
960
Length (Metric):
66 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Holds Length (Metric) Unit Size
10122862 Red, Green, Yellow, Blue 960 66 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10122862 Supplier Gilson™ Supplier No. F167100
  • ONLINE EXCLUSIVE PRICE
    21.15 EUR
    19.95 EUR / Pack of 1
    Save 1.20 EUR
    5% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
Required for use of Tower Pack tips. GilsonTM PIPETMANTM Universal Reload Box is color-coded.
  • TOWERPACKTM is an innovative, quick and easy refill system for DIAMONDTM Tips
  • Uses 60% less space on your laboratory bench and 70% less plastic
  • Entire range of DIAMONDTM Tips TOWERPACKTM fits the amazingly easy-to-use universal reload box
  • Color-coded racks differentiate tip models and indicate the compatible GilsonTM pipette models
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Red, Green, Yellow, Blue
Holds 960
Material Polypropylene
Positions 1
Shape Conical
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) Tower Pack
Quantity 1
Size 245 x 375 x 35 mm
Length (Metric) 66 mm
Length (English) 9.65 in
Width (English) 14.76 in
Height (English) 1.38 in
Includes Reload Box
Product Type Empty Rack
Array 1x1