Learn More
Gilson™ PIPETMANTM L Mechanical Pipet
GREENER_CHOICE
Product Code. p-4330400 Shop All Gilson Products
Change view
Click to view available options
Model:
P1000L
P100L
P10L
P10mL
P200L
P20L
P2L
P5000L
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
500 to 5000 μL
Accuracy:
±0.024, ±0.025, ±0.027, ±0.03μL
±0.025, ±0.075, ±0.1μL
±0.1, ±0.1, ±0.2μL
±0.35, ±0.4, ±0.8μL
±0.5, ±0.8, ±1.6μL
±12, ±15, ±30μL
±3, ±4, ±8μL
±30, ±40, ±60μL
Includes:
No Tip Ejector
Plastic Tip Ejector
Stainless Steel Tip Ejector
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Volume (Metric) Accuracy Includes Unit Size
12366142 P2L 0.2 to 2 μL ±0.024, ±0.025, ±0.027, ±0.03μL Stainless Steel Tip Ejector Each
12396142 P10L 1 to 10 μL ±0.025, ±0.075, ±0.1μL Plastic Tip Ejector Each
12306152 P20L 2 to 20 μL ±0.1, ±0.1, ±0.2μL Stainless Steel Tip Ejector Each
12376142 P2L 0.2 to 2 μL ±0.024, ±0.025, ±0.027, ±0.03μL Plastic Tip Ejector Each
12386142 P10L 1 to 10 μL ±0.025, ±0.075, ±0.1μL Stainless Steel Tip Ejector Each
12326152 P100L 10 to 100 μL ±0.35, ±0.4, ±0.8μL Stainless Steel Tip Ejector Each
12336152 P100L 10 to 100 μL ±0.35, ±0.4, ±0.8μL Plastic Tip Ejector Each
12346152 P200L 20 to 200 μL ±0.5, ±0.8, ±1.6μL Stainless Steel Tip Ejector Each
12356152 P200L 20 to 200 μL ±0.5, ±0.8, ±1.6μL Plastic Tip Ejector Each
12366152 P1000L 100 to 1000 μL ±3, ±4, ±8μL Stainless Steel Tip Ejector Each
12386152 P5000L 500 to 5000 μL ±12, ±15, ±30μL Stainless Steel Tip Ejector Each
12376152 P1000L 100 to 1000 μL ±3, ±4, ±8μL Plastic Tip Ejector Each
12396152 P10mL 1 to 10 mL ±30, ±40, ±60μL No Tip Ejector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 12366142 Supplier Gilson™ Supplier No. FA10001M
The PIPETMANTM L Mechanical Pipet is a lightweight pipet with lockable volume and low forces.
Mechanical pipette combines the legendary robustness of the PIPETMAN™ Classic with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Available in 8 single channel models covering the volume range of 0.2μL to 10mL.
  • Redesigned handle fits naturally in user's hand, no matter the size.
  • Volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • 2D identification code allows easy tracking of product information.
  • New piston assembly designed to drastically improve pipetting and purge forces.
  • Redesigned tip ejector shape and internal mechanism offers reduced tip ejection forces by 50%.
Order Info
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
TRUSTED_SUSTAINABILITY
Spezifikation
Accuracy ±0.024, ±0.025, ±0.027, ±0.03μL
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Orange
Dispensing Speed Manual
Disposable Reusable
For Use With D-10, DL10, DF10ST, DFL10ST Tip
Grip Type Ergonomic
Material Polypropylene
Model P2L
Number of Channels 1
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Mechanical Pipette
Volume (Metric) 0.2 to 2 μL
Finger Style Ergonomic
For Use With (Application) Transporting a measured volume of liquid
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Includes Stainless Steel Tip Ejector
Quantity 1
Warranty 1 Year
Length (English) 12.6 in.
Length (Metric) 32 cm
Width (English) 4.33 in.
Width (Metric) 11 cm
Height (English) 1.57 in.
Height (Metric) 4 cm
Fit for Purpose Certified Quality
Mehr anzeigen Weniger anzeigen