Learn More
Gilson™ PIPETMANTM Pipet
Product Code. 12366122 Shop All Gilson Products
Change view
Click to view available options
Color:
Blue
Orange
Purple
Red
Salmon
Yellow
Model:
P10
P100
P1000
P10mL
P2
P20
P200
P5000
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
500 to 5000 μL
Includes:
No ejector
Plastic Tip Ejector
Stainless Steel Tip Ejector
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Model Volume (Metric) Includes Unit Size
12366122 Red P10 1 to 10 μL Stainless Steel Tip Ejector Each
12346122 Orange P2 0.2 to 2 μL Stainless Steel Tip Ejector Each
11387790 Red P10 1 to 10 μL Plastic Tip Ejector Each
12356122 Orange P2 0.2 to 2 μL Plastic Tip Ejector Each
12386122 Yellow P20 2 to 20 μL Stainless Steel Tip Ejector Each
12396122 Yellow P20 2 to 20 μL Plastic Tip Ejector Each
12316132 Salmon P100 10 to 100 μL Plastic Tip Ejector Each
12306132 Salmon P100 10 to 100 μL Stainless Steel Tip Ejector Each
12326132 Yellow P200 20 to 200 μL Stainless Steel Tip Ejector Each
12366132 Purple P5000 500 to 5000 μL No ejector Each
12336132 Yellow P200 20 to 200 μL Plastic Tip Ejector Each
12346132 Blue P1000 100 to 1000 μL Stainless Steel Tip Ejector Each
12356132 Blue P1000 100 to 1000 μL Plastic Tip Ejector Each
12376132 Blue P10mL 1 to 10 mL No ejector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 12366122 Supplier Gilson™ Supplier No. F144055M
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Bulk Quote Request
PIPETMAN brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes with comfort.
  • Body (handle) in PVDF minimizes the hand-warming effect during pipetting and is resistant against aggressive chemicals
  • Direct volume reading in decimal digits helps prevent mistakes in volume setting
  • Comes with classic metal or plastic tip ejector
  • Clip ejector fits securely into the body of the pipette and helps avoid slipping over time
  • Redesigned seal, lighter ejection springs and high quality piston grease offer as low as 2 Newtons of force to pipet
  • High quality piston grease
Model Specific Features
  • Models P2 and P10 incorporate dual position tip ejectors for use with D10 (short) and DL10 (long) tips
Order Info
Trade in program available. Please contact your Fisher Scientific sales representative for more details.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±0.025, ±0.075, ±0.1μL
Compatible Tips D10, DL10, DF10ST, DFL10ST Tip
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Dispensing Speed Manual
Disposable Reusable
For Use With D-10, DL10, DF10ST, DFL10ST Tip
Grip Type Ergonomic
Material PVDF, Stainless Steel
Model P10
Number of Channels 1
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 1 to 10 μL
For Use With (Application) Extremely low pipetting forces, lightweight design
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Includes Stainless Steel Tip Ejector
Length (English) 14.37 in.
Length (Metric) 36.5 cm
Width (English) 12.01 in.
Width (Metric) 30.5 cm
Height (English) 4.72 in.
Height (Metric) 12 cm
Fit for Purpose Certified Quality