Learn More
Gibco™ CF1 Mouse Embryonic Fibroblasts, irradiated
Product Code. 15943412
Change view
Click to view available options
Quantity:
2 x 106 cells
4 x 106 cells
Unit Size:
Pack of 2
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15943412 4 x 106 cells Pack of 4
15815771 2 x 106 cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15943412 Supplier Gibco™ Supplier No. A34181
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Irradiated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) in culture. Manufactured by MTI-GlobalSt
  • Help save precious time with pre-inactivated, ready-to-use MEFs
  • Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
  • Rely on a widely-used product
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Cryopreserved mouse embryonic fibroblasts
Store in liquid nitrogen.
Age Embryonic
Cell Type Feeder Cells (Mouse Embryonic Fibroblast)
Form Cryopreserved
Species Mouse
Donor Source Pooled
Quantity 4 x 106 cells
Shipping Condition Shipped on dry ice