missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid, Potassium Salt 10%, MP Biomedicals™

Product Code. 11472960 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472960 5 g 5g
11482960 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472960 Supplier MP Biomedicals Supplier No. 0210028205

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Gibberellic acid is an endogenous plant growth regulator

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-67-7
Molecular Formula C19H21KO6
Molecular Weight (g/mol) 384.469
MDL Number MFCD00083342
InChI Key BGJIAUDTIACQSC-GLDAWBHVSA-M
Synonym unii-h52l7vzb7s, potassium gibberellate, gibberellic acid potassium salt, gibberellic acid, monopotassium salt, h52l7vzb7s, potassium gibberellin a3 1-, 77-06-5 parent, 2beta,7-dihydroxy-1-methyl-8-methylene-13-oxo-4aalpha,1alpha-epoxymethano gibb-3-ene-10beta-carboxylic acid potassium salt
PubChem CID 71299661
SMILES CC12C(C=CC3(C1C(C45C3CCC(C4)(C(=C)C5)O)C(=O)[O-])OC2=O)O.[K+]

Specifications

Assay Percent Range 10%
Quantity 5 g
Formula Weight 384.5
Physical Form Solid
Chemical Name or Material Gibberellic Acid, Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.