accessibility menu, dialog, popup

UserName

CAS RN 125-67-7

CH3CH2H2CHOOOOOHOK

CAS RN 125-67-7

IUPAC Name: potassium 5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylate
Synonyms: potassium 5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylate
Molecular Weight (g/mol): 384.47
Molecular Formula: C19H21KO6
InChi Key: BGJIAUDTIACQSC-UHFFFAOYNA-M
SMILES: [K+].CC12C3C(C([O-])=O)C45CC(=C)C(O)(C4)CCC5C3(OC1=O)C=CC2O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results