Learn More
Gibberellic Acid Methyl Ester Acetate Dimethyl Oxalate, TRC
Specifications
Specifications
| Chemical Name or Material | Gibberellic Acid Methyl Ester Acetate Dimethyl Oxalate |
| CAS | 102274-63-5 |
| Molecular Formula | C25 H28 O10 |
| Quantity | 100 mg |
| Synonym | (1alpha,2beta,4aalpha,4bbeta,10beta)-2-(Acetyloxy)-4a-hydroxy-7-[(methoxyoxoacetyl)oxy]-1-methyl-8-methylene-1,4a-lactone Gibb-3-ene-1,10-dicarboxylic Acid-10-methyl Ester |
| InChI Formula | InChI=1S/C25H28O10/c1-12-10-23-11-24(12,34-20(29)19(28)32-5)8-6-14(23)25-9-7-15(33-13(2)26)22(3,21(30)35-25)17(25)16(23)18(27)31-4/h7,9,14-17H,1,6,8,10-11H2,2-5H3/t14-,15+,16-,17-,22-,23+,24+,25-/m1/s1 |
| SMILES | COC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](OC(=O)C)C=C[C@@]2(OC3=O)[C@@H]4CC[C@@]5(C[C@]14CC5=C)OC(=O)C(=O)OC |
| IUPAC Name | 2-O-[(1R,2R,5S,8S,9S,10R,11S,12S)-12-acetyloxy-9-methoxycarbonyl-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.15,8.01,10.02,8]heptadec-13-en-5-yl] 1-O-methyl oxalate |
| Molecular Weight (g/mol) | 488.484 |
| Formula Weight | 488.168 |