Learn More
Brady™ GHS Symbol - Respiratory Hazard, Continuous Roll
Product Code. 15684227 Shop All Brady Products
Change view
Click to view available options
Material:
Polyester
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15684227 Polyester Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15684227 Supplier Brady™ Supplier No. 814029
  • ONLINE EXCLUSIVE PRICE
    409.00 EUR
    381.09 EUR / Pack of 250
    Save 27.91 EUR
    6% Off
View all promotions
In Stock
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1807-100*100-B7541-RLL-C
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Color Black on White