Learn More
Brady™ GHS Symbol - Oxidizing, Individual Markers on a Roll
Product Code. 15674827 Shop All Brady Products
Change view
Click to view available options
Material:
Polyester
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15674827 Polyester Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15674827 Supplier Brady™ Supplier No. 834180
  • ONLINE EXCLUSIVE PRICE
    58.60 EUR
    54.63 EUR / Pack of 250
    Save 3.97 EUR
    6% Off
View all promotions
In Stock
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1803-21*21-B7541-RLL-DC
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 21 mm
Thickness 0.099 mm
Width (Metric) 21 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Color Black on White