Learn More
Brady™ GHS Symbol - Explosive, Card Mounted
Product Code. 15484841 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
70 mm
100 mm
Width (Metric):
70 mm
100 mm
Unit Size:
Pack of 4
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
15484841 70 mm 70 mm Pack of 6
15454841 100 mm 100 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15484841 Supplier Brady™ Supplier No. 811682
  • ONLINE EXCLUSIVE PRICE
    14.95 EUR
    13.89 EUR / Pack of 6
    Save 1.06 EUR
    7% Off
Estimated Shipment: 04-12-2026
Pre-printed labels to identify your chemical product containers.
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Alternate Part Number PIC 1801-70*70-B7541-CRD
Finish Gloss
Includes 6 Labels/Card
Material Polyester
Quantity 6/Pk.
Adhesive Type Permanent Acrylic
Mounting Type Self-adhesive
For Use With (Application) Facility Identification, Hazard Communication Identification
Height (English) 2.76 in.
Height (Metric) 70 mm
Max. Temperature (English) 230°F
Max. Temperature (Metric) 110°C
Min. Temperature (English) -40°F
Thickness 0.099 mm
Width (English) 2.76 in.
Width (Metric) 70 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Temperature Range (English) -40°F to +230°F
Languages Universal
Product Type Hazard Sign
Color Red