Learn More
Thermo Scientific™ GFM Pro Gas Flowmeter
Product Code. 10744186 Shop All Thermo Scientific Products
Change view
Click to view available options
Airflow Rate:
0.5 to 500 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
10744186 0.5 to 500 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10744186 Supplier Thermo Scientific™ Supplier No. 66002010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
Estimated Shipment: 03-12-2026
Measure and monitor the flow of all types of gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter.
Measure and monitor the flow of all types of laboratoty gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter. Specifically designed for use with GC systems, this CE-certified flowmeter has a measurement range of 0.5–500mL/min and measures the volumetric flow of any gas with an accuracy of +/- 2% or +/- 0.2mL/min, whichever is greater. Its easy-to-use interface features an over-range warning indicator and automatic shutoff, and its explosion-proof rating allows use with flammable and explosive gases. The compact ergonomic design enables use as a hand-held or benchtop unit and features side grips for added durability.
Additional Product Information
  • Data output via USB port
  • Calibration: traceable to NIST primary standards
  • Uses two AA batteries
  • Recalibration service available
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±2% of Flow or ± 0.05 mL/min., whichever is greater
Airflow Rate 0.5 to 500 mL/min.
Description Electronic Gas Flowmeter