missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, 98%, MP Biomedicals™

Product Code. 11419601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419601 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419601 Supplier MP Biomedicals Supplier No. 0215718480

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 106-24-1
Color Colorless
Density 0.88g/mL
Assay Percent Range 0.98
Molecular Formula C10H18O
Quantity 100 mL
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
SMILES CC(C)=CCC\C(C)=C\CO
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular Weight (g/mol) 154.25
PubChem CID 637566
ChEBI CHEBI:17447
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material Geraniol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.