missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin C1 Pentaacetate Salt, TRC

Product Code. 17944385
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944385 1 mg 1mg
17904825 10 mg 10mg
17910665 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944385 Supplier Toronto Research Chemicals Supplier No. TRCG3605851MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25876-10-2
Molecular Formula C21H43N5O7.5C2H4O2
Molecular Weight (g/mol) 777.86
Synonym O-2-Amino-2,3,4,6,7-pentadeoxy-6-(methylamino)-α-D-ribo-heptopyranosyl-(14)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(16)]-2-deoxy-D-streptamine Pentaacetate Salt;
IUPAC Name acetic acid;(2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,6S)-3-amino-6-[(1R)-1-(methylamino)ethyl]tetrahydropyran-2-yl]oxy-2-hydroxy-cyclohexoxy]-5-methyl-4-(methylamino)tetrahydropyran-3,5-diol
SMILES CN[C@H](C)[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O

Specifications

Analyte or Component Names Gentamicin C1 Pentaacetate Salt
Shipping Condition Dry Pack
InChI Formula InChI=1S/C21H43N5O7.5C2H4O2/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;5*1-2(3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;5*1H3,(H,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;;;;;/m1...../s1
Solution Type Neat
Industry Type Forensics
Quantity 1 mg
Formula Weight 777.4219
Chemical Name or Material Gentamicin C1 Pentaacetate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.