CAS RN 25876-10-2
CAS RN 25876-10-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gentamicin C1 Pentaacetate Salt, TRC
CAS: 25876-10-2 Molecular Formula: C21H43N5O7.5C2H4O2 Molecular Weight (g/mol): 777.86 Synonym: O-2-Amino-2,3,4,6,7-pentadeoxy-6-(methylamino)-α-D-ribo-heptopyranosyl-(14)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(16)]-2-deoxy-D-streptamine Pentaacetate Salt; IUPAC Name: acetic acid;(2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,6S)-3-amino-6-[(1R)-1-(methylamino)ethyl]tetrahydropyran-2-yl]oxy-2-hydroxy-cyclohexoxy]-5-methyl-4-(methylamino)tetrahydropyran-3,5-diol SMILES: CN[C@H](C)[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O