Learn More
Thermo Scientific™ GENESYS Fiber Optic Probes
Product Code. 15841148 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometers
GENESYS 150/180 Spectrophotometer
Type:
Fiber Optic Coupler
Fiber Optic Dip Probe System
Fiber Optic Dip Probe, 10 mm
Tip for Dip Probe, 10 mm
Unit Size:
Each
Product Code. 15841148
Brand: Thermo Scientific™ 840305000
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Type Fiber Optic Dip Probe, 10 mm
For Use With (Equipment) GENESYS 150, GENESYS 180, Evolution One Plus, and Evolution Pro spectrophtometers