Learn More
3M™ General Purpose Masking Tape 201E
Product Code. 17833256 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
50 m x 24 mm
50 m x 48 mm
Unit Size:
Pack of 24
Pack of 36
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17833256 50 m x 24 mm Pack of 36
17843256 50 m x 48 mm Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17833256 Supplier 3M™ Supplier No. 7100195952
  • ONLINE EXCLUSIVE PRICE
    99.00 EUR
    83.72 EUR / Pack of 36
    Save 15.28 EUR
    15% Off
Estimated Shipment: 10-12-2026
The 3M™ General Purpose Masking Tape 201E is a premium-quality masking tape used for short-term paint masking, fixing, bundling, holding, sealing, and other applications up to a temperature of 80°C for one hour.
  • This tape is designed for general use in industrial applications with a slight need for temperature and water resistance such as short-term paint masking and sealing applications.
  • It provides a good instant adhesion and holding power that resists lifting or curling.
  • Its crepe paper backing offers conformability that will maintain integrity when formed around corners.
  • Its good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines due to its clean removability in one piece with no residue from most surfaces.
TRUSTED_SUSTAINABILITY
Specifications
Color Beige
Material Crepe Paper
Product Type Masking Tape
Dimensions (L x W) 50 m x 24 mm