Learn More
3M™ General Purpose Hazard Warning Tape 766
Product Code. 17711699 Shop All 3M Products
Change view
Click to view available options
Color:
Black with Yellow Stripes
Unit Size:
12 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17711699 Black with Yellow Stripes 12 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17711699 Supplier 3M™ Supplier No. 7100260599
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 01-12-2026
This General Purpose Hazard Warning Tape is ideal for the marking of equipment, pinch points, or gears for warning purposes. It can also be used for floor marking for light traffic areas, and around equipment or spills.
  • Ideal for safety marking applications in light traffic areas
  • Use for taping in all non-critical applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Applies instantly and is ready to use immediately
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Maintains vivid color over time for long-lasting quality and easy identification
  • Can be applied by hand or with floor applicator
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 33 m x 50 mm
Packaging Quantity 12 Rolls
Thickness 0.12 mm
Color Black with Yellow Stripes
Type Safety Tape