Learn More
Invitrogen™ GeneArt™ CRISPR Nuclease mRNA
Product Code. 15411054
Change view
Click to view available options
Quantity:
15 μg
Unit Size:
15µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411054 15 μg 15µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15411054 Supplier Invitrogen™ Supplier No. A29378
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Invitrogen™ GeneArt™ CRISPR Nuclease mRNA is formulated using a new manufacturing process to achieve superior quality and improved product performance.
  • This wild-type Cas9 mRNA has two nuclear localization signals (NLSs).
  • Cas9 constructs with two NLSs have been shown to be more efficient at targeting the Cas9 protein to the nucleus (1).
  • The mRNA format is ready to transfect and circumvents the need for time-consuming cloning steps required when using CRISPR vector systems.
  • Cas9 mRNA is co-transfected with a target-specific guide RNA (gRNA) to direct the Cas9 protein to the intended genome locus to create a double-stranded break.
  • The complete RNA format is a smaller payload than plasmid-based Cas9 systems for improved delivery into the cell and better genome editing efficiency.
  • Additionally, the Cas9 mRNA can be used in multiplex approaches with more than one gRNA.
  • Use this approach to determine which gRNA sequence works best for a particular target or to edit multiple genomic loci with one transfection.
TRUSTED_SUSTAINABILITY
Spécification
Content And Storage 1 vial CRISPR Nuclease mRNA. Store in ultra-cold freezer -68 to -85°C
Quantity 15 μg
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.