missing translation for 'onlineSavingsMsg'
Learn More

Gendenafil, TRC

Product Code. 30133118
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133118 25 mg 25mg
30006406 10 mg 10mg
30006412 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133118 Supplier Toronto Research Chemicals Supplier No. TRCG34996025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147676-66-2
Molecular Formula C19 H22 N4 O3
Molecular Weight (g/mol) 354.4
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-(5-acetyl-2-ethoxyphenyl)-1,6-dihydro-1-methyl-3-propyl-, 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-(5-acetyl-2-ethoxyphenyl)-1,4-dihydro-1-methyl-3-propyl- (9CI), 5-(5-Acetyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
IUPAC Name 5-(5-acetyl-2-ethoxyphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H22N4O3/c1-5-7-14-16-17(23(4)22-14)19(25)21-18(20-16)13-10-12(11(3)24)8-9-15(13)26-6-2/h8-10H,5-7H2,1-4H3,(H,20,21,25)
Formula Weight 354.1692
Percent Purity >95%
Chemical Name or Material Gendenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.