Learn More
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted
Product Code. 10788081
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10788081 25 mL Each
10000281 50 mL Each
10662662 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10788081 Supplier Lenz Laborglasintrumente™ Supplier No. 05630022
  • ONLINE EXCLUSIVE PRICE
    40.00 EUR
    31.15 EUR / Pack of 1
    Save 8.85 EUR
    22% Off
View all promotions
Estimated Shipment: 29-12-2026
Pycnometers acc. to Gay Lussac, unadjusted
  • Gay Lussac pycnometers acc. to DIN ISO 3507 and in addition to standard
  • Unadjusted
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Quantity 1
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)