Learn More
- Home
- Products
- Pumps
- Pump Parts and Accessories
- Pump Seals
- RETSCH Gasket For Grinding Jar
RETSCH Gasket For Grinding Jar
Product Code. 15350984 Shop All Retsch Products
Change view
Click to view available options
Material:
Stainless Steel
Stainless Steel, PTFE
Zirconium Oxide
Zirconium Oxide, Tungsten Carbide
For Use With (Equipment):
1.5 mL Grinding Jar
10 mL Grinding Jar
25 mL Grinding Jar
35 mL Grinding Jar
5 mL Grinding Jar
50 mL Grinding Jar
Quantity:
1 Pk.
10 Pk.
Unit Size:
Each
Pack of 10
13 product options available for selection
| Product Code. | Material | For Use With (Equipment) | Quantity | Unit Size |
|---|---|---|---|---|
| 15350984 | Stainless Steel, PTFE | 10 mL Grinding Jar | 1 Pk. | Each |
| 11924362 | Stainless Steel, PTFE | 1.5 mL Grinding Jar | 10 Pk. | Each |
| 10466739 | Stainless Steel, PTFE | 10 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 15330984 | Stainless Steel, PTFE | 25 mL Grinding Jar | 1 Pk. | Each |
| 15340984 | Zirconium Oxide | 25 mL Grinding Jar | 1 Pk. | Each |
| 15320984 | Stainless Steel, PTFE | 35 mL Grinding Jar | 1 Pk. | Each |
| 10250315 | Zirconium Oxide, Tungsten Carbide | 25 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 11944362 | Stainless Steel | 35 mL Grinding Jar | 10 Pk. | Each |
| 15310984 | Stainless Steel, PTFE | 50 mL Grinding Jar | 1 Pk. | Each |
| 10472080 | Zirconium Oxide | 35 mL Grinding Jar | 10 Pk. | Pack of 10 |
| 15300984 | Stainless Steel, PTFE | 5 mL Grinding Jar | 1 Pk. | Each |
| 11934362 | Stainless Steel, PTFE | 5 mL Grinding Jar | 10 Pk. | Each |
| 10290773 | Stainless Steel | 25 mL Grinding Jar | 10 Pk. | Pack of 10 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
This item is not returnable.
View return policy
Product Code.
15350984
Supplier
RETSCH
Supplier No.
03.111.0313
This item is not returnable.
View return policy
Specifications
Specifications
| For Use With (Equipment) | 10 mL Grinding Jar |
| Quantity | 1 Pk. |
| Material | Stainless Steel, PTFE |
| Type | Gasket |