Learn More
R&L Enterprises™ Wall Mounting Gas Cylinder Support
Product Code. 12995961 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Cylinders up to 230mm dia.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12995961 Cylinders up to 230mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 12995961
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Holds cylinders firmly and securely in a vertical position in the laboratory. R&L Enterprises™ Wall Mounting Gas Cylinder Supports are comprised of a curved nylon-coated steel bracket.
  • With plated chain
  • Suitable for use with cylinders up to 230mm in diameter
Specifications
Product Type Gas Cylinder Support Wall Fixing
For Use With (Equipment) Cylinders up to 230mm dia.