Learn More
R&L Enterprises™ Bench Mounting Gas Cylinder Support
Product Code. 10545784 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Cylinders up to 230mm dia.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10545784 Cylinders up to 230mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10545784
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Reduces danger of accidents from falling cylinders or escaping gases, up to 286mm in diameter. R&L Enterprises™ Bench Mounting Gas Cylinder Supports come with fitting instructions.
  • Incorporates two screw clamps and holes for permament fixing
  • Heavy metal bracket with fire-resistant strap and buckle
  • Secured permanently or temporarily to a bench edge up to 70mm thick
Specifications
Product Type Gas Cylinder Support Bench Mounting
For Use With (Equipment) Cylinders up to 230mm dia.