Learn More
Ansell GAMMEX Latex EnLite Gloves
Product Code. 17875473 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Unit Size:
50 pairs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17875473 6 50 pairs
17815483 5.5 50 pairs
17845473 6.5 50 pairs
17865473 7 50 pairs
17895473 7.5 50 pairs
17855473 8 50 pairs
17885473 8.5 50 pairs
17805483 9 50 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17875473 Supplier Ansell Supplier No. 330061060
GAMMEX Latex EnLite Gloves are powder-free reliable surgical gloves with a comfortable fit.
  • Unique anatomic formers enhances comfort
  • Exclusive manufacturing processes minimize the risk of developing allergies and sensitivities
  • A chlorination treatment helps make donning easy
  • Straight cuff provides a secure hold on the gown
  • Smooth with micro-textured finish for secure instrument handling and grip
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4, ISO 10282, JIS T9107
Accelerator Free No
Applications Healthcare
AQL 0.65
Color Natural
Disposable Reusable
Exterior Finish Smooth
Finger Thickness (English) 8.66 ± 0.79 in.
Finger Thickness (Metric) 0.22 ± 0.02 mm
Hand Specific
Laboratory/Cleanroom Use Laboratory
Latex Free No
Material Natural Rubber Latex
Palm Thickness (English) 8.07 ± 0.79 in.
Palm Thickness (Metric) 0.21 ± 0.02 mm
Powdered Powder Free
Product Type Surgical Gloves
Size 6
Sterility Sterile
Thickness (English) 8.07 ± 0.79 in.
Thickness (Metric) 0.21 ± 0.02 mm
Packaging Type Carton
Length (English) 11.2 in.
Length (Metric) 285 mm