Learn More
Gamma-Lindane-13C6, TRC
Product Code. 30472910
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472910 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472910 Supplier Toronto Research Chemicals Supplier No. TRCL4659920.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-HCH-13C6
CAS 104215-85-2
Molecular Formula 13C6 H6 Cl6
Quantity 0.5 mg
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane-13C6, gamma-HCH 13C6
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES Cl[13C@@H]1[13C@@H](Cl)[13C@@H](Cl)[13C@H](Cl)[13C@@H](Cl)[13C@H]1Cl
IUPAC Name 1,2,3,4,5,6-hexachloro(1,2,3,4,5,6-^{13}C_{6})cyclohexane
Molecular Weight (g/mol) 296.79
Formula Weight 293.8802