Learn More
WTW™ Galvanic Dissolved Oxygen Sensors (universal)
Product Code. 10769361
Change view
Click to view available options
Length (Metric) Cable:
1.5 m
3 m
6 m
10 m
15 m
Includes:
1 exchange membrane head
3 exchange membrane heads
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Includes Unit Size
10769361 1.5 m 3 exchange membrane heads Each
10503833 3 m 1 exchange membrane head Each
10235731 10 m 3 exchange membrane heads Each
10776701 15 m 3 exchange membrane heads Each
10470632 3 m 3 exchange membrane heads Each
10491592 6 m 3 exchange membrane heads Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10769361 Supplier WTW™ Supplier No. 201533
Estimated Shipment: 03-12-2026
Get accurate testing results for water and waterous solutions with these membrane-covered galvanic dissolved oxygen sensors that are low maintenance and no polarizarion time required.
  • Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
  • Can be used in the lab as well as in the field
  • Features include a quick response time (t99<60 s) and low-maintenance operation
  • Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
  • Filled with electrolyte and achieves a long dwelling time up to 6 months
  • Calibration vessel is part of the delivery
  • For use with conductivity and multiparameter meters
  • Includes Cable, Cleaning solution, abrasive foil, exchange membrane head(s) and electrolyte
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Cable 1.5 m
Includes 3 exchange membrane heads
For Use With (Equipment) Conductivity and multiparameter meters