missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product falls under category 12 PAP and aligns with the Green Chemistry Principle of Catalysis.
Specifications
Specifications
| CAS | 7440-55-3 |
| Melting Point | 29.8°C |
| Boiling Point | 2000°C |
| Density | 5.904 g/cm3 at 20°C |
| Quantity | 5 g |
| Form | Solid |
| MDL Number | MFCD00134045 |
| Synonym | Gallium, Gallium |
| Molecular Weight (g/mol) | 69.72 |
| Chemical Name or Material | Gallium |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?