Learn More
(-)-Galanthaminyl (-)-Camphanate, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Galanthaminyl (-)-Camphanate |
| CAS | 1025881-27-9 |
| Molecular Formula | C27 H33 N O6 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C27H33NO6/c1-24(2)25(3)10-11-27(24,34-22(25)29)23(30)32-17-8-9-26-12-13-28(4)15-16-6-7-18(31-5)21(20(16)26)33-19(26)14-17/h6-9,17,19H,10-15H2,1-5H3/t17-,19-,25-,26-,27+/m0/s1 |
| SMILES | COc1ccc2CN(C)CC[C@@]34C=C[C@@H](C[C@@H]3Oc1c24)OC(=O)[C@@]56CC[C@@](C)(C(=O)O5)C6(C)C |
| Molecular Weight (g/mol) | 467.55 |
| Formula Weight | 467.2308 |