Learn More
Gabazine, MedChemExpress
Product Code. 16473858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473858 10 mg 10mg
16446587 5 mg 5mg
16404896 100 mg 100mg
16400237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16473858 Supplier Medchem Express Supplier No. HY103533
 Restricted Item
Estimated Shipment: 09-11-2026
MedChemExpress Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.
Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ∼0.2 μM for GABA receptor.
Specifications
Chemical Name or Material Gabazine
CAS 104104-50-9
Color White
Purity Grade Notes Research
Molecular Formula C15H18BrN3O3
Quantity 10 mg
Synonym SR95531
Solubility Information H2O : 100 mg/mL (271.57 mM; Need ultrasonic) ∣DMSO : ≥ 75 mg/mL (203.68 mM)
SMILES O=C(O)CCCN1C(C=CC(C2=CC=C(OC)C=C2)=N1)=N.[H]Br
Molecular Weight (g/mol) 368.23
Formula Weight 368.23
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation