missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fusidic acid sodium salt, MedChemExpress
Description
Fusidic acid sodium salt (Sodium fusidate), a bacteriostatic antibiotic produced from the Fusidium coccineum fungus, belongs to the class of steroids. Fusidic acid sodium salt has no corticosteroid effects. Fusidic acid sodium salt inhibits the growth of bacteria by preventing the release of translation elongation factor G (EF-G) from the ribosome.
Specifications
Specifications
| Chemical Name or Material | Fusidic acid sodium salt |
| Physical Form | Solid |
| CAS | 751-94-0 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C31H47NaO6 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Sodium fusidate SQ-16360 |
| Solubility Information | H2O : ≥ 100 mg/mL (185.64 mM) ∣DMSO : ≥ 100 mg/mL (185.64 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?