Learn More
Furyl Hydroxymethyl Ketone, TRC
Product Code. 30134412
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134412 5 g 5g
30004752 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134412 Supplier Toronto Research Chemicals Supplier No. TRCF8652505G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17678-19-2
Molecular Formula C6 H6 O3
Molecular Weight (g/mol) 126.11
Synonym 2-Furyl Hydroxymethyl Ketone, 1-(2-Furanyl)-2-hydroxyethanone, 2-(1-Oxo-2-hydroxyethyl)furan, 2-(2'-Hydroxyacetyl)furan, 2-(Hydroxyacetyl)furan, 2-Furyl hydroxymethyl Ketone
IUPAC Name 1-(furan-2-yl)-2-hydroxyethanone
SMILES OCC(=O)c1occc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H6O3/c7-4-5(8)6-2-1-3-9-6/h1-3,7H,4H2
Formula Weight 126.0317
Percent Purity >95%
Chemical Name or Material Furyl Hydroxymethyl Ketone
Grade HPLC