missing translation for 'onlineSavingsMsg'
Learn More

Fursultiamine, TRC

Product Code. 30011756
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011756 25 mg 25mg
30007600 50 mg 50mg
30010999 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011756 Supplier Toronto Research Chemicals Supplier No. TRCF86523525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 804-30-8
Molecular Formula C17 H26 N4 O3 S2
Molecular Weight (g/mol) 398.54
Synonym Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-buten-1-yl]-, Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[(tetrahydrofurfuryl)dithio]-1-butenyl]- (6CI,8CI), Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-butenyl]- (9CI), N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-buten-1-yl]formamide, Adventan, Alinamin F, Diteftin, Fursultiamin, Fursultiamine, Judolor, Linamin, N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[(tetrahydrofurfuryl)dithio]-1-butenyl]formamide, Retar B1, TTFD, Tetrahydrofurfuryl thiamine disulfide, Thiamin tetrahydrofurfuryl disulfide, Thiamine tetrahydrofurfuryl disulfide
IUPAC Name N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[5-hydroxy-3-(oxolan-2-ylmethyldisulfanyl)pent-2-en-2-yl]formamide
SMILES CC(=C(CCO)SSCC1CCCO1)N(Cc2cnc(C)nc2N)C=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H26N4O3S2/c1-12(16(5-6-22)26-25-10-15-4-3-7-24-15)21(11-23)9-14-8-19-13(2)20-17(14)18/h8,11,15,22H,3-7,9-10H2,1-2H3,(H2,18,19,20)
Formula Weight 398.1446
Percent Purity >95%
Chemical Name or Material Fursultiamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.