Learn More
Carbolite™ Furnace with Uniform Temperature Envelope
Product Code. 12953741
Change view
Click to view available options
Capacity (Metric):
5 L
15 L
23 L
Temperature (Metric):
1200°C
1300°C
1600°C
Dimensions (D x W x H) Interior:
250 x 140 x 135 mm
305 x 220 x 220 mm
400 x 245 x 235 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Temperature (Metric) Dimensions (D x W x H) Interior unitSize
12953741 23 L 1300°C 400 x 245 x 235 mm Each
12183300 15 L 1600°C 305 x 220 x 220 mm Each
10704923 23 L 1200°C 400 x 245 x 235 mm Each
12933741 5 L 1300°C 250 x 140 x 135 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12953741 Supplier Carbolite™ Supplier No. CWF1323230SN+&02301
Tekniske data
Controller Type 301 PID Controller
Capacity (Metric) 23 L
Temperature (Metric) 1300°C
Type Furnace
Dimensions (D x W x H) Interior 400 x 245 x 235 mm
Width (Metric) 505 mm
Width (Metric) Interior 254 mm
Height (Metric) 705 mm
Height (Metric) Interior 235 mm
Depth (Metric) 675 mm
Depth (Metric) Interior 400 mm
Electrical Requirements 230 V 50/60 Hz 7000 w
Voltage 230 V
Wattage 7000 w
Frequency 50/60 Hz
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.