missing translation for 'onlineSavingsMsg'
Learn More

Furan-3-sulfonyl chloride, ≥97%, Thermo Scientific™

Product Code. p-3175299
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10620124 250 mg Amber Glass Bottle 250mg
10125773 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10620124 Supplier Thermo Scientific Maybridge Supplier No. CC02103CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 52665-49-3
Molecular Formula C4H3ClO3S
Molecular Weight (g/mol) 166.575
MDL Number MFCD08271875
InChI Key PCBBEIXTVWWTAK-UHFFFAOYSA-N
Synonym 3-furansulfonyl chloride, furan-3-sulfonylchloride, chloro-3-furylsulfone, 3-furansulfonylchlorid, 3-furansulfonylchloride, 3-furylsulfonyl chloride, 3-chlorosulphonyl furan, 3-furansulfonyl chloride;, 3-furanylsulfonyl chloride, 3-furylsulfonyl chloride;
PubChem CID 22179302
IUPAC Name furan-3-sulfonyl chloride
SMILES C1=COC=C1S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 166.58
Percent Purity 97%
Chemical Name or Material Furan-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.