Learn More
Furaltadone-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Furaltadone-d5 |
| CAS | 1015855-64-7 |
| Molecular Formula | C13 D5 H11 N4 O6 |
| Quantity | 50 mg |
| Synonym | 5-(4-Morpholinylmethyl-d2)-3-[[(5-nitro-2-furanyl)methylene]amino]-2-oxazolidinone-4,4,5-d3 |
| InChI Formula | InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2/b14-7+/i8D2,9D2,11D |
| SMILES | [2H]C([2H])(N1CCOCC1)C2([2H])OC(=O)N(\N=C\c3oc(cc3)[N+](=O)[O-])C2([2H])[2H] |
| IUPAC Name | 4,4,5-trideuterio-5-[dideuterio(morpholin-4-yl)methyl]-3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 329.32 |
| Formula Weight | 329.138 |