Learn More
asecos™ Funnel
Shop All asecos Products
Click to view available options
Material:
Stainless Steel
Steel
Unit Size:
Each
Description
- Funnel for easy filling of safety containers
Specifications
Specifications
| Product Type | Funnel |
| Color | Gray |
| For Use With (Application) | Pouring liquids |
| Material | Stainless Steel |
| Quantity | 1 |
| Width (English) | 4.33 in. |
| Width (Metric) | 110.00 mm |
| Height (English) | 9.06 in. |
| Height (Metric) | 230.00 mm |
| Weight (English) | 1.54 lb. |
| Show More |