missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 99.5% C{60}

Product Code. p-7050922
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304687 100 mg 100mg
11394677 1 g 1g
11384677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304687 Supplier Thermo Scientific Alfa Aesar Supplier No. 039722.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99685-96-8 | C60 | 720.66 g/mol

Fullerene powder is useful for biomedical applications such as the design of high-performance MRI contrast agents, x-ray imaging contrast agents and photodynamic therapy. It is also essential for single-walled carbon nanotubes with narrow diameter-distribution. It is used as light-activated antimicrobial agent. It also finds application in the field of nanotechnology, heat resistance and superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD00151408
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

Specifications

Color Black-Brown
Density 1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 100 mg
UN Number UN1325
Solubility Information Soluble in toluene. Insoluble in water.
Percent Purity 99.5%
Physical Form Powder
Assay C60
Chemical Name or Material Fullerene powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.