missing translation for 'onlineSavingsMsg'
Learn More

FTI 276 TFA, MedChemExpress

Product Code. 16463506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463506 25 mg 25mg
16452947 50 mg 50mg
16446888 10 mg 10mg
16429427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463506 Supplier Medchem Express Supplier No. HY15873A
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FTI-276 is a protein farnesyl transferase (PFT) inhibitor with IC50s of 0.9 nM and 0.5 nM for Plasmodium falciparum and human, respectively.

FTI-276 is a protein farnesyl transferase (PFT) inhibitor with IC50s of 0.9 nM and 0.5 nM for Plasmodium falciparum and human, respectively.

Specifications

Chemical Name or Material FTI 276 TFA
Physical Form Solid
CAS 1217471-51-6
Color Tan
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C23H28F3N3O5S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (182.61 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=CC=CC=C2)=O)CCSC.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 547.61
Formula Weight 547.61
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.