Learn More
Vileda Professional™ Sieve for Pre-Prepared Mop Box
Product Code. 17872466
Change view
Click to view available options
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17872466 Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17872466 Supplier Vileda Professional™ Supplier No. 120779
  • ONLINE EXCLUSIVE PRICE
    172.00 EUR
    163.39 EUR / Pack of 3
    Save 8.61 EUR
    5% Off
Estimated Shipment: 13-01-2027
For use with Pre-Mop Box
  • Fits on top of the Pre-Prepared Mop Box
  • Evenly placed holes which ensure even saturation of mops
  • Autoclavable
Specifications
Product Type Pre-Mop Box Sieve
For Use With (Equipment) Pre-Mop Box