Learn More
Gardner Denver Welch™ Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
Product Code. 15331461 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Rotary vane pump DN 16 KF
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15331461 Rotary vane pump DN 16 KF Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15331461 Supplier Gardner Denver Welch™ Supplier No. 330044
  • ONLINE EXCLUSIVE PRICE
    148.00 EUR
    146.30 EUR / Pack of 1
    Save 1.70 EUR
    1% Off
Estimated Shipment: 02-04-2027
For rotary vane pump DN 16 KF
Vacuum concentrator kit includes:
  • KF16 flange adapter
  • 1.5m PVC vacuum hose
  • Clamps, centring rings and adapters
TRUSTED_SUSTAINABILITY
Specifications
Description DN 16 KF
For Use With (Equipment) Rotary vane pump DN 16 KF