Learn More
Welch™ Freeze Dryer Adapter Kit 3
Product Code. 10084924 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Freeze Dryer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10084924 Freeze Dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10084924 Supplier Welch™ Supplier No. 330031
  • ONLINE EXCLUSIVE PRICE
    677.00 EUR
    559.10 EUR / Pack of 1
    Save 117.90 EUR
    17% Off
Estimated Shipment: 14-12-2026
This adapter kit is specifically designed for use with Welch™ Freeze Dryers and is used to connect various types of laboratory glassware to the freezer dryer system. The Kit 3 includes different adapters and connectors and is a convenient accessory to enhance versatility and functionality.
  • Allows connection of various types of laboratory glassware to the freeze dryer system
  • Connection: DN 16 KF
  • Includes different adapters and connectors allowing for easy attachment of flasks, vials, ampules or other glassware
  • PVC Construction
  • Enables efficient and controlled freeze drying of samples
  • Enhances versatility and functionality
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Freeze Dryer