Learn More
Gibco™ Freedom™ DG44 Kit
GREENER_CHOICE
Product Code. 11380812
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380812 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11380812 Supplier Gibco™ Supplier No. A1373701
Freedom™ DG44 Kit
Gibco™ Freedom™ DG44 Kit is an easy-to-use, beginning-to-end product, for cloning and expression of recombinant proteins in dihydrofolate reductase deficient (DHFR-) Chinese Hamster Ovary cells. The Gibco™ Freedom™ DG44 Kit includes components for transfection, expression, clone creation, and stable cell line selection, all in a conveniently packaged kit. The Gibco™ Freedom™ DG44 Kit also delivers a proven host cell line with simplified commercial licensing options.
  • Flexible commercial licensing without the need for royalties
  • Proven host used in approved products and a wide array of proteins
  • Complete workflow that can take your gene of interest from transfection to lead clone typically in less than 6 months
  • Convenient packaging, for simplified ordering and storage
    Commercial Licensing that is Simple, Flexible and Without Royalties
    Multiple companies provide CHO (Chinese Hamster Ovary) stable cell line development services and products that require annual fees during development of your therapeutic protein. In addition, royalties may be required on the sales of your therapeutic protein upon commercialization. Gibco™ Freedom™ Kits provide you with the tools that enable you or your CRO provider to develop your stable cell line for just the price of the kit itself. Furthermore, as you move towards commercial use, a flexible license structure is available without the burden of royalties and works to keep it as simple as a onetime payment (Fig 1). For more information regarding licensing, email us at outlicensing@lifetech.com.
    Host Cell Background with Proven Results
    DG44 cells have been a choice of scientists for decades for the expression of IgG and numerous blood proteins. Protein expression levels can vary significantly depending on the class of protein, however gram per liter levels of IgG expression has been demonstrated using these DG44 parental cells. So you can be confident that if your work calls for use of DG44 cells, they will deliver expected protein levels.
    Complete, Fast, and Efficient Workflow
    The Gibco™ Freedom™ DG44 Kit’s complete, proven, and fully integrated workflow allows you to move from transfection to lead clone typically in less then 6 months, helping to reduce hands on time and Fulltime Equivalent (FTE) requirements (Fig 2). We have implemented a single round of MTX (methotrexate) amplification in the protocol, which in our hands provided up to 120 fold titer improvement in less than 6 weeks (Fig 3). The entire workflow is performed in animal origin free (AOF) medium. The kit also offers the following advantages:
  • Cloning of one or two genes that encode your protein(s) of interest
  • High efficiency transfection of DNA into DG44 Cells (cGMP-banked)
  • Included flash drive with loaded manual
    Access to Gibco™ Freedom™ Kit Support
    If any specific support is needed beyond the manual, please contact your account manager to obtain a Process Science Manager consultation. In addition, one of our support scientists may check in with you to ensure that your experience with the kit is as successful as possible. For further assistance, please email our support team at GibcoServices@thermofisher.com.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage CD DG44 Medium: 2°C to 8°C, protect from light
CD OptiCHO™ Medium: 2°C to 8°C, protect from light
OptiPRO™ SFM: 2°C to 8°C, protect from light
FreeStyle™ MAX Reagent: 2°C to 8°C, do not freeze
Pluronic™: F68 15°C to 30°C
DG44 (CHO) cGMP Banked suspension dhfr (dihydrofolate reductase) Negative Cells: liquid nitrogen
One Shot™ TOP10 Chemically Competent E. coli: –80°C
pcDNA 3.3 TOPO™ TA Vector Kit: –5°C to –20°C
pOptiVEC™ TOPO™ TA Vector Kit: –5°C to –20°C
Gentamicin™: –5°C to –20°C
Product Line Freedom
Quantity 1 kit