missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ FRBH Boilersuit

Product Code. 31702828
Change view
Click to view available options
Size:
X-Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31702828 Medium Pack of 10
31702833 X-Small Pack of 10
31702824 Large Pack of 10
31702847 X-Large Pack of 10
31702831 2X-Large Pack of 10
31702843 3X-Large Pack of 10
31702845 4X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31702828 Supplier Skytec™ Supplier No. SKC030100EF
Add to basket
Add to basket
This item is not returnable. View return policy

Protective boilersuit designed to provide chemical resistance along with protection from heat, flame, and electrostatic hazards, while ensuring comfort for the wearer.

  • Offers protection against heat, flame and electrostatic hazards
  • Double storm facing velcro closure for increased protection
  • Ring-pull zip for easy donning and doffing when wearing gloves

Specifications

Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material PVC
Color Green
Disposable Reusable
Waist Style Loose
Ankle Style Elastic
Cuff Style Open
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals, Flame
Certifications/Compliance EN 1149-5:2018 or - EN 11612:2015 A1,B1,C1 or - EN 14605:2005 + A1:2009 Type 3,Type 4
For Use With (Application) Public Works, Transportation of Fuels, Waste Collection and Sorting, Chemical Handling and Containment
Weight (Metric) 1210 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.