Learn More
Tocris Bioscience™ FRAX 486
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
FRAX 486 is a potent p21-activated kinase (PAK) inhibitor (IC50 values are 14, 33, 39 and 575 nM for PAK1, PAK2, PAK3 and PAK4 respectively). Blocks and reverses the DISC1 knockdown-induced reduction in dendritic spine size in cortical neurons. Attenuates dendritic spine elimination and enhances spine generation in DISC1 knockdown mice. Ameliorates autism-like behavioral symptoms in fragile X mental retardation 1 (Fmr1) knockout mice. Brain penetrant and orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | FRAX 486 |
| Purity | >98% |
| Molecular Weight (g/mol) | 513.39 |
| Inhibitors | FRAX 486 |
For Research Use Only