Learn More
Gilson™ Fraction Collector FC204
Product Code. 16656145 Shop All Gilson Products
Change view
Click to view available options
Type:
With High Pressure Valve
With Low Pressure Valve
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
16656145 With High Pressure Valve Each
16646145 With Low Pressure Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16656145 Supplier Gilson™ Supplier No. 171043
  • ONLINE EXCLUSIVE PRICE
    9730.00 EUR
    8227.26 EUR / Pack of 1
    Save 1502.74 EUR
    15% Off
Estimated Shipment: 05-01-2027
The FC 204 Fraction Collector is a compact, large-bed fraction collector with a large capacity capable of handling test tubes, vials, and up to eight shallow or deep well microplates. Fractions can be collected by drop, peak, or time, and the collector can be used with any HPLC system.
  • Adaptable to Many Collection Strategies Collect samples by peak, drop, or time using the FC 204 Fraction Collector.
  • Add up to ten programmable time windows to collect only what you want, eliminating void volumes, equilibration volumes, and peaks of no interest.
  • In peak mode, the collector monitors a detector signal, identifying peaks using an adaptive-slope algorithm or a specified millivolt threshold.
  • The 3-way diverter valve prevents contamination of fractions. Multicolumn adapters allow for collection from up to 18 separate channels.
  • Easy to Operate With the easy-to-use keypad, setup and standalone operation are simplified. The collector can also be controlled through a PC using the TRILUTION™ LC software.
  • Using contact signals, the FC 204 can be connected to any HPLC system.
Specifications
Type With High Pressure Valve
Max. Flow Rate 200 mL/min
Voltage 110/220 V