missing translation for 'onlineSavingsMsg'
Learn More

Fosthiazate, TRC

Product Code. 30041073
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041073 50 mg 50mg
30040715 1 g 1g
30004295 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041073 Supplier Toronto Research Chemicals Supplier No. TRCF74580050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98886-44-3
Molecular Formula C9 H18 N O3 P S2
Molecular Weight (g/mol) 283.35
Synonym Phosphonothioic acid, (2-oxo-3-thiazolidinyl)-, O-ethyl S-(1-methylpropyl) ester (9CI), Fosthiazate, IKI 1145, Nemathorin, S-sec-Butyl O-ethyl (2-oxo-3-thiazolidinyl)phosphonothioate, S-sec-Butyl O-ethyl (2-oxo-3-thiazolidinyl)phosphonothiolate
IUPAC Name 3-[butan-2-ylsulfanyl(ethoxy)phosphoryl]-1,3-thiazolidin-2-one
SMILES CCOP(=O)(SC(C)CC)N1CCSC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H18NO3PS2/c1-4-8(3)16-14(12,13-5-2)10-6-7-15-9(10)11/h8H,4-7H2,1-3H3
Formula Weight 283.0466
Percent Purity >95%
Chemical Name or Material Fosthiazate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.