Learn More
Fostamatinib Disodium, MedChemExpress
Product Code. 16498217
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498217 5 mg 5mg
16456086 100 mg 100mg
16455508 10 mg 10mg
16411687 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16498217 Supplier Medchem Express Supplier No. HY13038
 Restricted Item
Estimated Shipment: 07-10-2026
MedChemExpress Fostamatinib Disodium (R788 Disodium) is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
Fostamatinib Disodium (R788 Disodium) is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).
Specifications
Chemical Name or Material Fostamatinib Disodium
CAS 1025687-58-4
Color White
Purity Grade Notes Research
Molecular Formula C23H24FN6Na2O9P
Quantity 5 mg
Synonym R788(Disodium)
Solubility Information DMSO : 50 mg/mL (80.07 mM; Need ultrasonic)
SMILES COC1=CC(NC2=NC=C(C(NC3=NC(N4COP(O[Na])(O[Na])=O)=C(C=C3)OC(C)(C)C4=O)=N2)F)=CC(OC)=C1OC
Molecular Weight (g/mol) 624.42
Formula Weight 624.42
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation